Merck
Z744854
Squib Separatory Funnel with Stopcock
new
capacity 300 mL, borosilicate glass
Sign In to View Organizational & Contract Pricing
Select a Size
About This Item
UNSPSC Code:
41121809
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
PTFE stopcock
borosilicate glass
Joint
Joints stopcock (4)
joint stopper24/40
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 1426T-300
capacity
300 mL
General description
The Squibb Style Separatory Funnel with stopper features a smooth tooled tip, large white block letters, and large marking squares.
Features and Benefits
  • Supplied with PTFE stopcocks to eliminate freezing and lubricant contamination
  • Made from high-quality borosilicate tubing with a low coefficient of expansion
  • Stopper No.: 24/40
  • Stopcock Plug Size: 4
Choose from one of the most recent versions:
Certificates of Analysis (COA)