Merck
  • Comparison of the helix-coil transition of a titrating polypeptide in aqueous solutions and at the air-water interface.
Comparison of the helix-coil transition of a titrating polypeptide in aqueous solutions and at the air-water interface.
Biophysical chemistry (2005-05-25)
Helen Sjögren, Stefan Ulvenlund
ABSTRACT
The transition from alpha-helix to random coil of the titrating polyamino acid co-poly-L-(lysine, phenylalanine), (p-(Lys,Phe)), has been investigated as a function of pH and ionic strength in aqueous solution and at the air-water interface by means of circular dichroism (CD) spectroscopy and the Langmuir surface film balance technique. The results strongly suggest that the helix-coil transition for peptides at the air-water interface can be determined by using the two-dimensional Flory exponent, nu, to express the pH dependent peptide surface conformation. The helix-coil titration curve of p-(Lys,Phe) shifts approximately 2.5 pH units towards lower pH at the air-water interface, as compared with the bulk solution. This finding is of relevance for the understanding of conformation and conformational changes of membrane-transporting and membrane penetrating peptides as well as for the use of peptides in molecular devices.
Sigma-Aldrich
Poly(Lys, Phe) 1:1 hydrobromide, mol wt 20,000-50,000
Pricing and availability is not currently available.