Merck
  • The magnesium transporter NIPAL1 is a pancreatic islet-expressed protein that conditionally impacts insulin secretion.
The magnesium transporter NIPAL1 is a pancreatic islet-expressed protein that conditionally impacts insulin secretion.
The Journal of biological chemistry (2020-05-23)
Yousef Manialawy, Saifur R Khan, Alpana Bhattacharjee, Michael B Wheeler
ABSTRACT
Type 2 diabetes is a chronic metabolic disease characterized by pancreatic β-cell dysfunction and peripheral insulin resistance. Among individuals with type 2 diabetes, ∼30% exhibit hypomagnesemia. Hypomagnesemia has been linked to insulin resistance through reduced tyrosine kinase activity of the insulin receptor; however, its impact on pancreatic β-cell function is unknown. In this study, through analysis of several single-cell RNA-sequencing data sets in tandem with quantitative PCR validation in both murine and human islets, we identified NIPAL1 (NIPA-like domain containing 1), encoding a magnesium influx transporter, as an islet-enriched gene. A series of immunofluorescence experiments confirmed NIPAL1's magnesium-dependent expression and that it specifically localizes to the Golgi in Min6-K8 cells, a pancreatic β-cell-like cell line (mouse insulinoma 6 clone K8). Under varying magnesium concentrations, NIPAL1 knockdown decreased both basal insulin secretion and total insulin content; in contrast, its overexpression increased total insulin content. Although the expression, distribution, and magnesium responsiveness of NIPAL1 in α-TC6 glucagonoma cells (a pancreatic α-cell line) were similar to the observations in Min6-K8 cells, no effect was observed on glucagon secretion in α-TC6 cells under the conditions studied. Overall, these results suggest that NIPAL1 expression is regulated by extracellular magnesium and that down-regulation of this transporter decreases glucose-stimulated insulin secretion and intracellular insulin content, particularly under conditions of hypomagnesemia.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Glucagon EIA Kit, for serum, plasma, culture supernatant and cell lysates