Merck
Structural Basis of Functional Transitions in Mammalian NMDA Receptors.
Cell (2020-07-02)
Tsung-Han Chou, Nami Tajima, Annabel Romero-Hernandez, Hiro Furukawa
ABSTRACT
Excitatory neurotransmission meditated by glutamate receptors including N-methyl-D-aspartate receptors (NMDARs) is pivotal to brain development and function. NMDARs are heterotetramers composed of GluN1 and GluN2 subunits, which bind glycine and glutamate, respectively, to activate their ion channels. Despite importance in brain physiology, the precise mechanisms by which activation and inhibition occur via subunit-specific binding of agonists and antagonists remain largely unknown. Here, we show the detailed patterns of conformational changes and inter-subunit and -domain reorientation leading to agonist-gating and subunit-dependent competitive inhibition by providing multiple structures in distinct ligand states at 4 Å or better. The structures reveal that activation and competitive inhibition by both GluN1 and GluN2 antagonists occur by controlling the tension of the linker between the ligand-binding domain and the transmembrane ion channel of the GluN2 subunit. Our results provide detailed mechanistic insights into NMDAR pharmacology, activation, and inhibition, which are fundamental to the brain physiology.
Product Description
Millipore
Digitonin, High Purity, CAS 11024-24-1 is a non-ionic detergent commonly used to solubilize membrane-bound proteins.
Pricing and availability is not currently available.