Merck
  • Neuroprotective effect of ketamine against TNF-α-induced necroptosis in hippocampal neurons.
Neuroprotective effect of ketamine against TNF-α-induced necroptosis in hippocampal neurons.
Journal of cellular and molecular medicine (2021-03-05)
Lu Wang, Bin Deng, Panpan Yan, Huanghui Wu, Chunhui Li, Hongrui Zhu, Jiwei Du, Lichao Hou
ABSTRACT
Tumour necrosis factor-α (TNF-α), a crucial cytokine, has various homeostatic and pathogenic bioactivities. The aim of this study was to assess the neuroprotective effect of ketamine against TNF-α-induced motor dysfunction and neuronal necroptosis in male C57BL/6J mice in vivo and HT-22 cell lines in vitro. The behavioural testing results of the present study indicate that ketamine ameliorated TNF-α-induced neurological dysfunction. Moreover, immunohistochemical staining results showed that TNF-α-induced brain dysfunction was caused by necroptosis and microglial activation, which could be attenuated by ketamine pre-treatment inhibiting reactive oxygen species production and mixed lineage kinase domain-like phosphorylation in hippocampal neurons. Therefore, we concluded that ketamine may have neuroprotective effects as a potent inhibitor of necroptosis, which provides a new theoretical and experimental basis for the application of ketamine in TNF-α-induced necroptosis-associated diseases.
Sigma-Aldrich
Fluorometric Intracellular Ros Kit, sufficient for 200 fluorometric tests (green)
Pricing and availability is not currently available.