Merck
  • Analysis of single- and double-stranded nucleic acids on polyacrylamide and agarose gels by using glyoxal and acridine orange.
Analysis of single- and double-stranded nucleic acids on polyacrylamide and agarose gels by using glyoxal and acridine orange.
Proceedings of the National Academy of Sciences of the United States of America (1977-11-01)
G K McMaster, G G Carmichael
PMID73185
ABSTRACT
We have developed a simple and rapid system for the denaturation of nucleic acids and their subsequent analysis by gel electrophoresis. RNA and DNA are denatured in 1 M glyoxal (ethanedial) and 50% (vol/vol) dimethyl sulfoxide, at 50 degrees. The glyoxalated nucleic acids are then subjected to electrophoresis through either acrylamide or agarose gels in a 10 mM sodium phosphate buffer at pH 7.0. When glyoxalated DNA molecules of known molecular weights are used as standards, accurate molecular weights for RNA are obtained. Furthermore, we have employed the metachromatic stain acridine orange for visualization of nucleic acids in gels. This dye interacts differently with double- and single-stranded polynucleotides, fluorescing green and red, respectively. By using these techniques, native and denatured DNA and RNA molecules can be analyzed on the same slab gel.
Sigma-Aldrich
Acridine Orange hemi(zinc chloride) salt, For nucleic acid staining in cells or gels
Pricing and availability is not currently available.