Merck
  • Determination of flavonoids from Orthosiphon stamineus in plasma using a simple HPLC method with ultraviolet detection.
Determination of flavonoids from Orthosiphon stamineus in plasma using a simple HPLC method with ultraviolet detection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-01-25)
Yit Hong Loon, Jia Woei Wong, Siew Ping Yap, Kah Hay Yuen
ABSTRACT
A simple liquid chromatographic method was developed for the simultaneous determination of flavonoids from Orthosiphon stamineus Benth, namely sinensitin, eupatorin and 3'-hydroxy-5,6,7,4'-tetramethoxyflavone, in plasma. Prior to analysis, the flavonoids and the internal standard (naproxen) were extracted from plasma samples using a 1:1 mixture of ethyl acetate and chloroform. The detection and quantification limits for the three flavonoids were similar being 3 and 5 ng/ml, respectively. The within-day and between-day accuracy values, expressed as percentage of true values, for the three flavonoids were between 95 and 107%, while the corresponding precision, expressed as coefficients of variation, for the three flavonoids were less than 14%. In addition, the mean recovery values of the extraction procedure for all the flavonoids were between 92 and 114%. The calibration curves were linear over a concentration range of 5-4000 ng/ml. The present method was applied to analyse plasma samples obtained from a pilot study using rats in which the mean absolute oral bioavailability values for sinensitin, eupatorin and 3'-hydroxy-5,6,7,4'-tetramethoxyflavone was 9.4, 1.0 and 1.5%, respectively.
Sigma-Aldrich
Eupatorin, ≥97% (HPLC)
Pricing and availability is not currently available.