Merck
  • PS-COD and PS-9-BBN: polymer-supported reagents for solution-phase parallel synthesis.
PS-COD and PS-9-BBN: polymer-supported reagents for solution-phase parallel synthesis.
Organic letters (2005-02-25)
Jefferson D Revell, Barbara Dörner, Peter D White, A Ganesan
ABSTRACT
1,5-Cyclooctadiene was deprotonated under LICKOR conditions and reacted with Merrifield resin to afford an immobilized cyclooctadiene in high yield. This polymer is effective as a halogen scavenger, while hydroboration leads to a supported 9-BBN analogue. The latter exhibits similar regioselectivity to 9-BBN in olefin hydroboration. [reaction: see text]
Sigma-Aldrich
9-Borabicyclo[3.3.1]nonane solution, 0.4 M in hexanes
Pricing and availability is not currently available.
Sigma-Aldrich
9-Borabicyclo[3.3.1]nonane solution, 0.5 M in THF
Pricing and availability is not currently available.