Merck
  • Binding of epidermal growth factor-dextran conjugates to cultured glioma cells.
Binding of epidermal growth factor-dextran conjugates to cultured glioma cells.
International journal of cancer (1991-02-01)
A Andersson, A Holmberg, J Carlsson, J Carlsson, J Pontén, B Westermark
ABSTRACT
Some gliomas, melanomas and squamous carcinomas have large numbers of EGF receptors which could, in these cases, be used for targeting with toxic agents. We investigated whether EGF could be conjugated to dextran, which is a suitable carrier for toxic agents, without losing its ability to bind to the receptor. Dextran of 20 kDa molecular weight was activated with I-cyano-4-dimethylamino pyridinium tetrafluoroborate (CDAP) yielding highly active pyridinium-isourea derivatives. EGF was coupled to the activated dextran through the amino terminus and glycine was added to block residual activity. The EGF-dextran conjugate was, after purification on Sephadex G25 and Sephacryl 200 columns, tested for its receptor binding properties on human malignant glioma, U343MGaC12:6, cells. The conjugate inhibited binding of 125I-EGF in a competitive assay, showing that the binding was receptor-specific. Dextran conjugated with glycine, without EGF, had no inhibitory effect. The conjugate was radio-labelled either on the EGF part with 125I or on the dextran part with 3H-glycine, and the internalization patterns were compared to the internalization of 125I-EGF. The radioactivity of the conjugates remained cell-associated for more than 20 hr, regardless of whether the radioactivity was on the EGF or on the dextran part, while the radioactivity of unconjugated EGF rapidly disappeared from the cells. Most of the cell-associated radioactivity was, at all analysed time intervals, located intracellularly. Thus, it seems promising to use dextran, conjugated with EGF, as a carrier of, for example, toxic radioactive nuclides.
Sigma-Aldrich
1-Cyano-4-dimethylaminopyridinium tetrafluoroborate, organic cyanylating reagent
Pricing and availability is not currently available.