Merck
  • Role of vinylcatechin in the formation of pyranomalvidin-3-glucoside-+-catechin.
Role of vinylcatechin in the formation of pyranomalvidin-3-glucoside-+-catechin.
Journal of agricultural and food chemistry (2008-10-31)
Luis Cruz, Natércia Teixeira, Artur M S Silva, Nuno Mateus, Jose Borges, Victor de Freitas
ABSTRACT
Reactions between malvidin-3-glucoside (mv3glc) and 8-vinylcatechin were carried out to synthesize pyranomv3glc-(+)-catechin pigment and to study the formation of intermediates. A rapid decrease of mv3glc content concomitant with the formation of more complex structures such as mv3glc-vinylcatechin [precursor of pyranomv3glc-(+)-catechin pigment] and mv3glc-divinylcatechin was observed. On the other hand, 8-vinylcatechin undergoes acid-catalyzed dimerization in model wine solution, giving rise to 8-vinylcatechin dimers. These compounds were also found in the reaction between mv3glc and (+)-catechin mediated by acetaldehyde, which provides evidence for the formation of 8-vinylcatechin and its involvement in the formation of pyranoanthocyanins in aged red wines.
Sigma-Aldrich
Oenin chloride, ≥90% (HPLC)
Pricing and availability is not currently available.