Merck
  • Inhibitory effects of albuterol and fenoterol on RANTES and IP-10 expression in bronchial epithelial cells.
Inhibitory effects of albuterol and fenoterol on RANTES and IP-10 expression in bronchial epithelial cells.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology (2011-02-16)
Ka-Pan Lam, Yu-Te Chu, Min-Sheng Lee, Huan-Nan Chen, Wei-Li Wang, Teck-Siang Tok, Yow-Yue Chin, Solomon Chih-Cheng Chen, Chang-Hung Kuo, Chih-Hsing Hung
ABSTRACT
Short-acting β2-adrenoreceptor agonist (SABA) is the major asthma reliever as indicated in the GINA guidelines. Regulated on activation, normal T expressed and secreted (RANTES) is a chemokine that attracts eosinophils, mast cells, and basophils toward site of allergic inflammation. Interferon γ-inducible protein (IP)-10 is a Th1-related chemokine that is also important in asthmatic inflammation and also involved in our immune defense against pathogens. Bronchial epithelial cells are first-line barrier against invasive pathogen and also have immunomodulatory function. However, whether albuterol and fenoterol (two SABAs) have modulatory effects on RANTES and IP-10 expression in bronchial epithelial cells is unknown. The human bronchial epithelial cell lines, BEAS-2B cells, were pre-treated with different concentrations of albuterol, fenoterol or dibutyryl-cAMP (a cyclic AMP analog) before polyinosinic-polycytidylic acid (poly I:C) stimulation. In some condition, BEAS-2B cells were pre-treated with ICI-118551, a selective β2-adrenoreceptor antagonist, 30 min before albuterol or fenoterol treatment. The levels of RANTES and IP-10 were measured by ELISA. Intracellular signaling was investigated using cAMP assay, mitogen-activated protein kinase (MAPK) inhibitor, nuclear factor (NF)-κB inhibitor, and western blot. Albuterol and fenoterol suppressed poly I:C-induced RANTES and IP-10 expression of BEAS-2B cells. ICI-118551 could partly reverse the suppressive effects of albuterol and fenoterol on RANTES and IP-10 expression. Albuterol and fenoterol increased intracellular cAMP levels. Dibutyryl-cAMP conferred the similar effects of albuterol and fenoterol. Western blot revealed that albuterol suppressed p-ERK, p-JNK and pp38, and also their associated kinase expression. Albuterol had no effect on pp65 expression. Albuterol and fenoterol could suppress poly I:C-induced RANTES and IP-10 expression in human bronchial epithelial cells via at least partly the β2-adrenoreceptor-cAMP and the MAPK pathways, implicating that albuterol and fenoterol could exert anti-inflammatory effect and benefit asthmatic patients by suppressing RANTES and IP-10 expression. However, these suppressive effects of albuterol and fenoterol may inhibit the defense against viral infection.
Sigma-Aldrich
Fenoterol hydrobromide
Pricing and availability is not currently available.