- Efficient synthesis of β-enaminones and β-enaminoesters catalyzed by gold (I)/silver (I) under solvent-free conditions.
Efficient synthesis of β-enaminones and β-enaminoesters catalyzed by gold (I)/silver (I) under solvent-free conditions.
Molecules (Basel, Switzerland) (2012-03-08)
Ming Zhang, Ablimit Abdukader, Yong Fu, Chengjian Zhu
PMID22395409
ABSTRACT
An efficient method for the synthesis of β-enaminones and β-enaminoesters using a combination of [(PPh(3))AuCl]/AgOTf as catalyst has been developed. The reaction between 1,3-dicarbonyl compounds and primary amines was carried out under solvent-free conditions with low catalyst loading in good to excellent yields at room temperature.