Merck
  • Continuous biodegradation of parathion by immobilized Sphingomonas sp. in magnetically fixed-bed bioreactors and evaluation of the enzyme stability of immobilized bacteria.
Continuous biodegradation of parathion by immobilized Sphingomonas sp. in magnetically fixed-bed bioreactors and evaluation of the enzyme stability of immobilized bacteria.
Biotechnology letters (2012-09-22)
Seyed Mortaza Robatjazi, Seyed Abbas Shojaosadati, Rassoul Khalilzadeh, Ebrahim Vasheghani Farahani, Mehdi Zeinoddini
ABSTRACT
Magnetically-modified Sphingomonas sp. was prepared using covalent binding of magnetic nanoparticles on to the cell surface. The magnetic modified bacteria were immobilized in the fixed-bed bioreactors (FBR) by internal and external magnetic fields for the biodetoxification of a model organophosphate, parathion: 93 % of substrate (50 mg parathion/l) was hydrolyzed at 0.5 ml/min in internal magnetic field fixed-bed bioreactor. The deactivation rate constants (at 1 ml/min) were 0.97 × 10(-3), 1.24 × 10(-3) and 4.17 × 10(-3) h(-1) for immobilized bacteria in external and internal magnetic field fixed-bed bioreactor and FBR, respectively. The deactivation rate constant for immobilized magnetically modified bacteria in external magnetic field fixed-bed bioreactor (EMFFBR) was 77 % lower than that of immobilized cells by entrapping method on porous basalt beads in FBR at 1 ml/min. Immobilized magnetic modified bacteria exhibited maximum enzyme stability in EMFFBR.
Product Description
Supelco
Parathion solution, 100 μg/mL in cyclohexane, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Parathion, PESTANAL®, analytical standard
Pricing and availability is not currently available.