- Further studies on oligoribonucleotide synthesis.
Further studies on oligoribonucleotide synthesis.
Nucleic acids symposium series (1980-01-01)
W T Markiewicz, E Biała, R W Adamiak, K Grześkowiak, R Kierzek, A Kraszewski, J Stawiński, M Wiewiórowski
PMID7255163
ABSTRACT
Recent results concerning the synthesis of oligoribonucleotides via the phosphotriester method, such as functionalization of ribonucleosides, new phosphorylating agents, 5'-O-sulfonylation and chromatography on Sephadex LH-20 for monitoring the removal of internucleotide phosphotriester groups, are presented. To show that efficiency of a new approach to the synthesis of oligoribonucleotides the pentamer /Up/4U was obtained.