Merck
Fragrance material review on alpha-amylcinnamyl alcohol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-27)
S P Bhatia, G A Wellington, J Cocchiara, J Lalko, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of alpha-amylcinnamyl alcohol when used as a fragrance ingredient is presented.
Product Description
Supelco
α-Amylcinnamyl alcohol, analytical standard
Pricing and availability is not currently available.