Merck
Sulfur K-edge photofragmentation of ethylene sulfide.
The Journal of chemical physics (2010-07-10)
W C Stolte, G Ohrwall
ABSTRACT
We have investigated the photofragmentation properties of the three-membered ring heterocyclic molecule ethylene sulfide or thiirane, C(2)H(4)S, by time-of-flight mass spectroscopy. Positive ions have been collected as a function of photon energy around the S K ionization threshold. Branching ratios were derived for all detected ions, which are informative of the decay dynamics and photofragmentation patterns of the core-excited species. We present a new assignment of the spectral features around the S K-edge.
Sigma-Aldrich
Ethylene sulfide, 98%
Pricing and availability is not currently available.