Merck
  • N-WASP coordinates the delivery and F-actin-mediated capture of MT1-MMP at invasive pseudopods.
N-WASP coordinates the delivery and F-actin-mediated capture of MT1-MMP at invasive pseudopods.
The Journal of cell biology (2012-10-24)
Xinzi Yu, Tobias Zech, Laura McDonald, Esther Garcia Gonzalez, Ang Li, Iain Macpherson, Juliane P Schwarz, Heather Spence, Kinga Futó, Paul Timpson, Colin Nixon, Yafeng Ma, Ines M Anton, Balázs Visegrády, Robert H Insall, Karin Oien, Karen Blyth, Jim C Norman, Laura M Machesky
ABSTRACT
Metastasizing tumor cells use matrix metalloproteases, such as the transmembrane collagenase MT1-MMP, together with actin-based protrusions, to break through extracellular matrix barriers and migrate in dense matrix. Here we show that the actin nucleation-promoting protein N-WASP (Neural Wiskott-Aldrich syndrome protein) is up-regulated in breast cancer, and has a pivotal role in mediating the assembly of elongated pseudopodia that are instrumental in matrix degradation. Although a role for N-WASP in invadopodia was known, we now show how N-WASP regulates invasive protrusion in 3D matrices. In actively invading cells, N-WASP promoted trafficking of MT1-MMP into invasive pseudopodia, primarily from late endosomes, from which it was delivered to the plasma membrane. Upon MT1-MMP's arrival at the plasma membrane in pseudopodia, N-WASP stabilized MT1-MMP via direct tethering of its cytoplasmic tail to F-actin. Thus, N-WASP is crucial for extension of invasive pseudopods into which MT1-MMP traffics and for providing the correct cytoskeletal framework to couple matrix remodeling with protrusive invasion.
Sigma-Aldrich
Anti-MMP-14 Antibody, catalytic domain, clone LEM-2/15.8, clone LEM-2/15.8, Chemicon®, from mouse
Pricing and availability is not currently available.