Merck
  • The first total synthesis of kwakhurin, a characteristic component of a rejuvenating plant, "kwao keur": toward an efficient synthetic route to phytoestrogenic isoflavones.
The first total synthesis of kwakhurin, a characteristic component of a rejuvenating plant, "kwao keur": toward an efficient synthetic route to phytoestrogenic isoflavones.
Organic & biomolecular chemistry (2005-02-11)
Fumihiro Ito, Misako Iwasaki, Toshiko Watanabe, Tsutomu Ishikawa, Yoshihiro Higuchi
ABSTRACT
A convergent synthesis of kwakhurin (5), a characteristic estrogen-like isoflavone of Pueraria mirifica(Leguminosae), is described. Isoflavone skeleton 31 was constructed by Suzuki-Miyaura coupling of 3-bromochromone 26 (AC-ring) and arylboronic acid 30 (B-ring) in the presence of TBAB as an additive. Microwave-assisted coupling was also examined, but did not improve the yield. Baeyer-Villiger oxidation, followed by propargylation and reduction afforded 1,1-dimethylallyl ether 37. 6'-Prenylisoflavone 34 was obtained in high yield by Claisen rearrangement of 37 in N,N-diethylaniline. On the other hand, 1,3-rearrangement of prenyl ether 33 with clay gave 34 in poor yield. Successive methylation of 34 and deprotection yielded the target kwakhurin (5) in 12% overall yield from 2,4-dihydroxybenzaldehyde (23).
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Pricing and availability is not currently available.