Merck
  • Hbo1 is a cyclin E/CDK2 substrate that enriches breast cancer stem-like cells.
Hbo1 is a cyclin E/CDK2 substrate that enriches breast cancer stem-like cells.
Cancer research (2013-08-21)
Mylinh T Duong, Said Akli, Sira Macalou, Anna Biernacka, Bisrat G Debeb, Min Yi, Kelly K Hunt, Khandan Keyomarsi
ABSTRACT
Expression of cyclin E proteolytic cleavage products, low-molecular weight cyclin E (LMW-E), is associated with poor clinical outcome in patients with breast cancer and it enhances tumorigenecity in mouse models. Here we report that LMW-E expression in human mammary epithelial cells induces an epithelial-to-mesenchymal transition phenotype, increases the CD44(hi)/CD24(lo) population, enhances mammosphere formation, and upregulates aldehyde dehydrogenase expression and activity. We also report that breast tumors expressing LMW-E have a higher proportion of CD44(hi)/CD24(lo) tumor cells as compared with tumors expressing only full-length cyclin E. In order to explore how LMW-E enriches cancer stem cells in breast tumors, we conducted a protein microarray analysis that identified the histone acetyltransferase (HAT) Hbo1 as a novel cyclin E/CDK2 substrate. The LMW-E/CDK2 complex phosphorylated Hbo1 at T88 without affecting its HAT activity. When coexpressed with LMW-E/CDK2, wild-type Hbo1 promoted enrichment of cancer stem-like cells (CSC), whereas the T88 Hbo1 mutant reversed the CSC phenotype. Finally, doxorubicin and salinomycin (a CSC-selective cytotoxic agent) synergized to kill cells expressing LMW-E, but not full-length cyclin E. Collectively, our results suggest that the heightened oncogenecity of LMW-E relates to its ability to promote CSC properties, supporting the design of therapeutic strategies to target this unique function.
Sigma-Aldrich
Salinomycin, from Streptomyces albus, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Salinomycin monosodium salt, VETRANAL®, analytical standard
Pricing and availability is not currently available.