Merck
  • Aminocarbonyloxymethyl ester prodrugs of flufenamic acid and diclofenac: suppressing the rearrangement pathway in aqueous media.
Aminocarbonyloxymethyl ester prodrugs of flufenamic acid and diclofenac: suppressing the rearrangement pathway in aqueous media.
Archiv der Pharmazie (2007-01-09)
Lina Ribeiro, Nuno Silva, Jim Iley, Jarkko Rautio, Tomi Järvinen, Hélder Mota-Filipe, Rui Moreira, Eduarda Mendes
ABSTRACT
Aminocarbonyloxymethyl ester prodrugs are known to undergo rearrangement in aqueous solutions to form the corresponding N-acylamine side product via an O-->N intramolecular acyl transfer from the carbamate conjugate base. Novel aminocarbonyloxymethyl esters of diclofenac and flufenamic acid containing amino acid amide carriers were synthesized and evaluated as potential prodrugs displaying less ability to undergo rearrangement. These compounds were prepared in reasonable yield by a four-step synthetic method that uses the appropriate N-Boc-protected amino acid N-hydroxysuccinimide ester and secondary amine and chloromethyl chloroformate as key reactants. Their reactivity in pH 7.4 buffer and 80% human plasma at 37 degrees C was assessed by RP-HPLC. The aminocarbonyloxymethyl esters containing a secondary carbamate group derived from amino acids such as glycine or phenylalanine were hydrolyzed quantitatively to the parent drug both in non-enzymatic and enzymatic conditions, with no rearrangement product being detected. The oral bioavailability in rats was determined for selected diclofenac derivatives. These derivatives displayed a bioavailability of 25 to 68% relative to that of diclofenac, probably due to their poor aqueous solubility and lipophilicity. These results suggest that further optimization of aminocarbonyloxymethyl esters as potential prodrugs for non-steroidal anti-inflammatory drugs require the use of amino acid carriers with ionizable groups to improve aqueous solubility.
Sigma-Aldrich
Chloromethyl chloroformate, ≥98.0% (GC)
Pricing and availability is not currently available.