Merck
  • Dormant bacteria within Staphylococcus epidermidis biofilms have low inflammatory properties and maintain tolerance to vancomycin and penicillin after entering planktonic growth.
Dormant bacteria within Staphylococcus epidermidis biofilms have low inflammatory properties and maintain tolerance to vancomycin and penicillin after entering planktonic growth.
Journal of medical microbiology (2014-07-24)
Filipe Cerca, Angela França, Begoña Pérez-Cabezas, Virgínia Carvalhais, Adília Ribeiro, Joana Azeredo, Gerald Pier, Nuno Cerca, Manuel Vilanova
ABSTRACT
Staphylococcus epidermidis is the most commonly isolated aetiological agent of nosocomial infections, mainly due to its ability to establish biofilms on indwelling medical devices. Detachment of bacteria from S. epidermidis biofilms and subsequent growth in the planktonic form is a hallmark of the pathogenesis of these infections leading to dissemination. Here we showed that S. epidermidis cells collected from biofilms cultured in conditions that promote cell viability present marked changes in their physiological status upon initiating a planktonic mode of growth. When compared to cells growing in biofilms, they displayed an increased SYBR green I staining intensity, increased transcription of the rpiA gene, decreased transcription of the icaA gene, as well as higher susceptibility to vancomycin and penicillin. When bacteria collected from biofilms with high proportions of dormant cells were subsequently cultured in the planktonic mode, a large proportion of cells maintained a low SYBR green I staining intensity and increased resistance to vancomycin and penicillin, a profile typical of dormant cells. This phenotype further associated with a decreased ability of these biofilm-derived cells to induce the production of pro-inflammatory cytokines by bone marrow-derived dendritic cells in vitro. These results demonstrated that cells detached from the biofilm maintain a dormant cell-like phenotype, having a low pro-inflammatory effect and decreased susceptibility to antibiotics, suggesting these cells may contribute to the recalcitrant nature of biofilm infections.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Vancomycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Millipore
Vancomycin supplement, suitable for microbiology
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, meets USP testing specifications
SAFC
L-Glutamine
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥85% (Vancomycin B)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Magnesium chloride, ≥98%