- The hooded crow (Corvus cornix) as an environmental bioindicator species of heavy metal contamination.
The hooded crow (Corvus cornix) as an environmental bioindicator species of heavy metal contamination.
Bulletin of environmental contamination and toxicology (2014-08-26)
Mauro Giammarino, Piero Quatto, Stefania Squadrone, Maria Cesarina Abete
PMID25149280
ABSTRACT
This study aims to examine the possible presence of lead and cadmium in the liver and kidneys of hooded crows (Corvus cornix). Liver and kidneys of hooded crow carcasses were collected in Province of Cuneo (Piedmont, Italy) in order to detect lead and cadmium content. Significant differences were found in lead and cadmium levels between areas of intensive cultivation versus areas where meadows are prevalent. Moreover, age greatly influenced the burden of heavy metals, while sex did not seem to affect the level of contamination. The source of contamination may be phosphate fertilizers used for intensive cultivation in the study area.
Product Description
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Pricing and availability is not currently available.
Lead, foil, 15mm disks, thickness 0.03mm, 99.99+%
Pricing and availability is not currently available.
Cadmium, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95%
Pricing and availability is not currently available.
Cadmium, wire reel, 2m, diameter 0.5mm, hard, 99.99+%
Pricing and availability is not currently available.
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Pricing and availability is not currently available.
Lead, foil, not light tested, 150x150mm, thickness 0.05mm, 99.99+%
Pricing and availability is not currently available.
Cadmium, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Pricing and availability is not currently available.
Lead, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Pricing and availability is not currently available.
Lead, rod, 200mm, diameter 50mm, 99.95%
Pricing and availability is not currently available.
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.95+%
Pricing and availability is not currently available.
Lead, microfoil, disks, 10mm, thinness 0.25μm, specific density 266.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Pricing and availability is not currently available.
Lead, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.99+%
Pricing and availability is not currently available.
Lead, foil, not light tested, 300x300mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Pricing and availability is not currently available.
Lead, tube, 500mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Pricing and availability is not currently available.
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%
Pricing and availability is not currently available.
Lead, foil, light tested, 25x25mm, thickness 0.025mm, 99.99+%
Pricing and availability is not currently available.
Lead, wire reel, 0.5m, diameter 2.0mm, as drawn, 99.99+%
Pricing and availability is not currently available.
Lead, rod, 200mm, diameter 6.35mm, 99.95%
Pricing and availability is not currently available.
Lead, foil, not light tested, 50x50mm, thickness 0.03mm, 99.99+%
Pricing and availability is not currently available.
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Pricing and availability is not currently available.
Cadmium, foil, 50x200mm, thickness 1.0mm, as rolled, 99.999%
Pricing and availability is not currently available.
Lead, wire reel, 2m, diameter 3.0mm, as drawn, 99.99+%
Pricing and availability is not currently available.
Lead, foil, not light tested, 50x50mm, thickness 0.015mm, 99.99+%
Pricing and availability is not currently available.
Lead, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Pricing and availability is not currently available.
Cadmium, wire reel, 1m, diameter 0.5mm, hard, 99.95+%
Pricing and availability is not currently available.