Merck
Analysis of fentanyl in urine by DLLME-GC-MS.
Journal of analytical toxicology (2014-12-11)
Michael A Gardner, Sheena Sampsel, Werner W Jenkins, Janel E Owens
ABSTRACT
Fentanyl is a synthetic narcotic anesthetic ∼80-100 times more potent than morphine. Owing to the potential for its abuse, the drug may be included in a forensic toxicology work-up, which requires fast, precise and accurate measurements. Here, the stability of fentanyl was assessed when stored at three different temperatures (-20, 4 and 25°C) in synthetic urine. Stability at those three temperatures was demonstrated over 12 weeks upon analysis by gas chromatography-mass spectrometry with a deuterated internal standard (fentanyl-D5) utilizing three different extraction techniques: liquid-liquid extraction (LLE), solid-phase extraction and dispersed liquid-liquid microextraction (DLLME). The DLLME method was then optimized before use in the analysis of fentanyl in urine samples obtained from autopsy cases at the El Paso County Coroner's Office. Accuracy of the DLLME method was assessed by completing spike and recovery studies at three different fortification levels (10, 100 and 250 ng/mL) with excellent recovery (89.9-102.6%). The excellent comparability between DLLME and LLE is demonstrated (Bland-Altman difference plot with a mean difference of 4.9 ng/mL) and the use of this methodology in the analysis of forensically relevant samples is discussed.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
1-Chlorobutane, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
1-Chlorobutane, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chlorobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%