Merck
  • Selective extraction of metal ions from aqueous phase to ionic liquids: a novel thermodynamic approach to separations.
Selective extraction of metal ions from aqueous phase to ionic liquids: a novel thermodynamic approach to separations.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-08-27)
Camiel H C Janssen, Antonio Sánchez, Mark N Kobrak
ABSTRACT
The selective extraction of metals from aqueous mixtures has generally relied on the use of selective ionophores. We present an alternative strategy that exploits a recently developed approach to extraction into an ionic liquid phase, and show that a high degree of control over selectivity can be obtained by tuning the relative concentrations of extraction agents. A thermodynamic model for the approach is presented, and an experimental separation of strontium and potassium ions is performed. It is shown that tuning the concentrations of the species involved can shift the ratio of potassium to strontium in the ionic liquid phase from 4:1 to 3:4. This extraction is performed under mild conditions with relatively common reagents. The result is a proof-of-concept for a novel separations scheme that could have great importance in a wide range of technological applications.
Sigma-Aldrich
Dicyclohexano-18-crown-6, 98%
Pricing and availability is not currently available.