Merck
  • Role of SIRT1 in Streptococcus pneumoniae-induced human β-defensin-2 and interleukin-8 expression in A549 cell.
Role of SIRT1 in Streptococcus pneumoniae-induced human β-defensin-2 and interleukin-8 expression in A549 cell.
Molecular and cellular biochemistry (2014-06-05)
Li Lin, Shun-hang Wen, Shu-zhen Guo, Xiao-yan Su, Hu-jun Wu, Lei Chong, Hai-lin Zhang, Wei-xi Zhang, Chang-chong Li
ABSTRACT
Streptococcus pneumoniae is an important pathogen of pneumonia in human. Human alveolar epithelium acts as an effective barrier and is an active participant in host defense against invasion of bacterial by production of various mediators. Sirtuin 1 (SIRT1), the prototypic class III histone deacetylase, is involved in the molecular control of lifespans and immune responses. This study aimed at examining the role of SIRT1 in mediating S. pneumoniae-induced human β-defensin-2 (hBD2) and interleukin-8(IL-8) expression in the alveolar epithelial cell line A549 and the underlying mechanisms involved. A549 cells were infected with S. pneumoniae for indicated times. Exposure of A549 cells to S. pneumoniae increased the expressions of SIRT1 protein, hBD2 and IL-8 mRNA, and protein. The SIRT1 activator resveratrol enhanced S. pneumoniae-induced gene expression of hBD2 but decreased IL-8 mRNA levels. Blockade of SIRT1 activity by the SIRT1 inhibitors nicotinamide reduced S. pneumoniae-induced hBD2 mRNA expression but increased its stimulatory effects on IL-8 mRNA. S. pneumoniae-induced activation of extracellular signal-regulated kinase (ERK) and p38 mitogen-activated protein kinase (MAPK). SIRT1 expression was attenuated by selective inhibitors of ERK and p38 MAPK. The hBD2 mRNA production was decreased by pretreatment with p38 MAPK inhibitor but not with ERK inhibitor, whereas the IL-8 mRNA expression was controlled by phosphorylation of ERK. These results suggest that SIRT1 mediates the induction of hBD2 and IL-8 gene expression levels in A549 cell by S. pneumoniae. SIRT1 may play a key role in host immune and defense response in A549.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Resveratrol, European Pharmacopoeia (EP) Reference Standard