Merck
  • Metformin alleviates hepatosteatosis by restoring SIRT1-mediated autophagy induction via an AMP-activated protein kinase-independent pathway.
Metformin alleviates hepatosteatosis by restoring SIRT1-mediated autophagy induction via an AMP-activated protein kinase-independent pathway.
Autophagy (2014-12-09)
Young Mi Song, Yong-ho Lee, Ji-Won Kim, Dong-Sik Ham, Eun-Seok Kang, Bong Soo Cha, Hyun Chul Lee, Byung-Wan Lee
ABSTRACT
Metformin activates both PRKA and SIRT1. Furthermore, autophagy is induced by either the PRKA-MTOR-ULK1 or SIRT1-FOXO signaling pathways. We aimed to elucidate the mechanism by which metformin alleviates hepatosteatosis by examining the molecular interplay between SIRT1, PRKA, and autophagy. ob/ob mice were divided into 3 groups: one with ad libitum feeding of a standard chow diet, one with 300 mg/kg intraperitoneal metformin injections, and one with 3 g/d caloric restriction (CR) for a period of 4 wk. Primary hepatocytes or HepG2 cells were treated with oleic acid (OA) plus high glucose in the absence or presence of metformin. Both CR and metformin significantly improved body weight and glucose homeostasis, along with hepatic steatosis, in ob/ob mice. Furthermore, CR and metformin both upregulated SIRT1 expression and also stimulated autophagy induction and flux in vivo. Metformin also prevented OA with high glucose-induced suppression of both SIRT1 expression and SIRT1-dependent activation of autophagy machinery, thereby alleviating intracellular lipid accumulation in vitro. Interestingly, metformin treatment upregulated SIRT1 expression and activated PRKA even after siRNA-mediated knockdown of PRKAA1/2 and SIRT1, respectively. Taken together, these results suggest that metformin alleviates hepatic steatosis through PRKA-independent, SIRT1-mediated effects on the autophagy machinery.
Product Description
Supelco
Oleic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Epoxy embedding medium, accelerator, ≥95% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution
Pricing and availability is not currently available.
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® siRNA Universal Negative Control #1
Pricing and availability is not currently available.
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Oleic acid, technical grade, 90%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium chloride
Pricing and availability is not currently available.
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Pricing and availability is not currently available.
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Pricing and availability is not currently available.
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Pricing and availability is not currently available.
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Oleic acid, natural, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LDN-57444, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Oleic acid, Selectophore, ≥99%
Pricing and availability is not currently available.
Roche
DNase I, grade II, from bovine pancreas
Pricing and availability is not currently available.