Merck
  • A simian-human immunodeficiency virus carrying the rt gene from Chinese CRF01_AE strain of HIV is sensitive to nucleoside reverse transcriptase inhibitors and has a highly genetic stability in vivo.
A simian-human immunodeficiency virus carrying the rt gene from Chinese CRF01_AE strain of HIV is sensitive to nucleoside reverse transcriptase inhibitors and has a highly genetic stability in vivo.
Microbes and infection (2014-04-09)
Wei Wang, Nan Yao, Bin Ju, Zhihui Dong, Zhe Cong, Hong Jiang, Chuan Qin, Qiang Wei
ABSTRACT
Human immunodeficiency virus (HIV)-1 subtype CRF01_AE is one of the major HIV-1 subtypes that dominate the global epidemic. However, its drug resistance, associated mutations, and viral fitness have not been systemically studied, because available chimeric simian-HIVs (SHIVs) usually express the HIV-1 reverse transcriptase (rt) gene of subtype B HIV-1, which is different from subtype CRF01_AE HIV-1. In this study, a recombinant plasmid, pRT-SHIV/AE, was constructed to generate a chimeric RT-SHIV/AE by replacing the rt gene of simian immunodeficiency virus (SIVmac239) with the counterpart of Chinese HIV-1 subtype CRF01_AE. The infectivity, replication capacity, co-receptor tropism, drug sensitivity, and genetic stability of RT-SHIV/AE were characterized. The new chimeric RT-SHIV/AE effectively infected and replicated in human T cell line and rhesus peripheral blood mononuclear cells (rhPBMC). The rt gene of RT-SHIV/AE lacked the common mutation (T215I) associated with drug resistance. RT-SHIV-AE retained infectivity and immunogenicity, similar to that of its counterpart RT-SHIV/TC virus following intravenous inoculation in Chinese rhesus macaque. RT-SHIV-AE was more sensitive to nucleoside reverse transcriptase inhibitors (NRTIs) than the RT-SHIV/TC. RT-SHIV/AE was genetically stable in Chinese rhesus macaque. The new chimeric RT-SHIV/AE may be a valuable tool for evaluating the efficacy of the rt-based antiviral drugs against the subtype CRF01_AE HIV-1.
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%
SAFC
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Supelco
Aucubin, analytical standard
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethanolamine, ≥99%
Aucubin, primary reference standard
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, ≥98%