Merck
  • Faecal Escherichia coli from patients with E. coli urinary tract infection and healthy controls who have never had a urinary tract infection.
Faecal Escherichia coli from patients with E. coli urinary tract infection and healthy controls who have never had a urinary tract infection.
Journal of medical microbiology (2014-01-28)
Karen L Nielsen, Pia Dynesen, Preben Larsen, Niels Frimodt-Møller
ABSTRACT
Urinary tract infections (UTIs) are primarily caused by Escherichia coli with the patient's own faecal flora acting as a reservoir for the infecting E. coli. Here we sought to characterize the E. coli faecal flora of UTI patients and healthy controls who had never had a UTI. Up to 20 E. coli colonies from each rectal swab were random amplified polymorphic DNA (RAPD) typed for clonality, dominance in the sample and correlation to the infecting UTI isolate in patients. Each distinct clone was phylotyped and tested for antimicrobial susceptibility. Eighty-seven per cent of the UTI patients carried the infecting strain in their faecal flora, and faecal clones causing UTI were more often dominant in the faecal flora. Patients had a larger diversity of E. coli in their gut flora by carrying more unique E. coli clones compared to controls, and patient faecal clones were more often associated with multidrug resistance compared to controls. We found a similar phylotype distribution of faecal clones from UTI patients and healthy controls, including a large proportion of B2 isolates in the control group. Faecal-UTI isolates from patients were more often associated with multidrug resistance compared to faecal-only clones, indicating a link between UTI virulence and antimicrobial resistance. Intake of any antibiotic less than 6 months prior to inclusion in the experiment occurred significantly more in patients with UTI than in controls. In contrast, presence of an intrauterine device was significantly more common in controls indicating a protective effect against UTI. In conclusion, healthy controls have a large proportion of potentially pathogenic E. coli phylotypes in their faecal flora without this causing infection.
Product Description
Supelco
Mecillinam, VETRANAL®, analytical standard
Pricing and availability is not currently available.