Merck
  • No evidence of a role for cystatin B gene in juvenile myoclonic epilepsy.
No evidence of a role for cystatin B gene in juvenile myoclonic epilepsy.
Epilepsia (2015-03-11)
Laura Mumoli, Patrizia Tarantino, Roberto Michelucci, Amedeo Bianchi, Angelo Labate, Silvana Franceschetti, Carla Marini, Pasquale Striano, Monica Gagliardi, Edoardo Ferlazzo, Vito Sofia, Loredana Pennese, Grazia Annesi, Umberto Aguglia, Renzo Guerrini, Federico Zara, Antonio Gambardella
ABSTRACT
Genetic factors play a major role in the etiology of juvenile myoclonic epilepsy (JME), a common form of idiopathic generalized epilepsy, but so far, genes related to JME remain largely unknown. JME shares electroclinical features with Unverricht-Lundborg disease (progressive myoclonic epilepsy type 1; EPM1), a form of progressive myoclonus epilepsy characterized by myoclonus, epilepsy, and gradual neurologic deterioration. EPM1 is caused by mutations in the gene that codes for cystatin B (CSTB), an inhibitor of cysteine protease. In the present study, we wished to investigate the role of the CSTB gene in patients with JME. Fifty-seven unrelated patients (35 women; mean age ± standard deviation [SD], 24.1 ± 7.7; mean age ± SD at onset, 15.3 ± 2.4) with JME were enrolled. Twenty-three of 57 patients were the probands of families with JME. The molecular diagnosis was carried out to identify the common dodecamer repeat expansion mutation or other disease-causing mutations in the CSTB gene. The molecular analysis did not depict mutations in any of the 57 patients with JME. Our study did not support a role for the CSTB gene in patients with familial or sporadic JME.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®