Merck
  • Metabolomic analysis of lung epithelial secretions in rats: an investigation of bronchoalveolar lavage fluid by GC-MS and FT-IR.
Metabolomic analysis of lung epithelial secretions in rats: an investigation of bronchoalveolar lavage fluid by GC-MS and FT-IR.
Experimental lung research (2014-08-26)
Wajhul Qamar, Syed Rizwan Ahamad, Raisuddin Ali, Mohammad Rashid Khan, Abdul Rahman Al-Ghadeer
ABSTRACT
Rat bronchoalveolar lavage fluid (BALF) metabolome can be used to obtain valuable, precise, and accurate information about underlying lung conditions in an experiment. The present study focuses on the evaluation of the lung epithelium metabolome in a rat model using techniques including bronchoalveolar lavage, gas chromatography-mass spectroscopy (GC-MS), and Fourier transform infrared spectroscopy (FT-IR). Untargeted metabolites in BALF were extracted in ethyl acetate and derivatized by standard methods for the analysis by GC-MS. FT-IR spectra of ethyl acetate extract of BALF were obtained and read for the characteristic fingerprint of rats under investigation. Analyses were done in individual animals to obtain consistent data. BALF cells were counted by flow cytometry to monitor any inflammatory condition in rats. FT-IR analysis finds two peaks which are characteristically different from the extract medium, which is ethyl acetate. FT-IR peaks correspond to that of amino acids and carbohydrates, including β-D-glucose, α-D-glucose, and β-D-galactose. GC-MS evaluation of the BALF finds several products of the metabolism or its participants. Main compounds in the BALF detected by GC-MS include succinate, fumarate, glycine, alanine, 2-methyl-3-oxovaleric acid, dodecanoic acid, tetradecanoic acid, hexadecanoic acid, octanoic acid, trans-9-octadecanoic acid, octadecanoic acid, and Prostaglandin F1α. Several research reports reveal metabolomic parameters in murine model lung tissue or BALF, but they rarely reported a complete metabolomics model profile, particularly in rats. The present data of GC-MS and FT-IR suggest that the set up can be exploited to study metabolomic alterations in several lung conditions including acute lung toxicity, inflammation, asthma, bronchitis, fibrosis, and emphysema.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Ethyl acetate, analytical standard
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%