Merck
Biodistribution of size-selected lyophilisomes in mice.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-05-09)
Etienne van Bracht, René Raavé, Igor Y Perevyazko, Elly M Versteeg, Theo G Hafmans, Ulrich S Schubert, Egbert Oosterwijk, Toin H van Kuppevelt, Willeke F Daamen
ABSTRACT
Lyophilisomes are a novel class of proteinaceous biodegradable nano/microparticle capsules developed for tumor drug delivery. The in vivo characteristics of lyophilisomes are unknown and, therefore, the time course of biodistribution of sized albumin-based lyophilisomes in CD1 mice after intravenous administration was studied. Lyophilisomes, prepared from Dylight680-labeled albumin, were sized using a sucrose gradient centrifugation methodology and four fractions with a mean size of approximately 200nm, 400nm, 550nm, and 650nm were pooled for in/ex vivo localization, (immuno)histochemistry and biochemical analysis. Lyophilisomes were rapidly taken out of the circulation by the liver and spleen. Immunohistochemistry revealed that lyophilisomes were taken up in the liver by F4/80 positive macrophages, and in the spleen by Sign-R1 positive macrophages specifically located in the marginal zones. Lyophilisomes were most likely degraded by the liver and spleen and subsequently excreted via the urine, as high levels of degraded Dylight680-labeled albumin were detected in the urine. This was corroborated by electron microscopy of the spleen, which showed intact lyophilisomes in the marginal zone 5 and 30min after injection, but not after 2h. In conclusion, IV injected lyophilisomes are rapidly entrapped by liver and splenic macrophages, biodegraded, and excreted in the urine.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
SAFC
Glycine
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles