Merck
  • Experimental TLR4 inhibition improves intestinal microcirculation in endotoxemic rats.
Experimental TLR4 inhibition improves intestinal microcirculation in endotoxemic rats.
Microvascular research (2015-06-28)
Juan Zhou, Marieke Soltow, Katrin Zimmermann, Dragan Pavlovic, Brent Johnston, Christian Lehmann
ABSTRACT
Toll like receptor 4 (TLR4) represents a critical cellular link for endotoxin-induced pathology. The aim of this study was to evaluate the potential role of TLR4 inhibition on the intestinal microcirculation during experimental endotoxemia. The intestinal microcirculation was studied by intravital microscopy in four groups of Lewis rats (n=10 per group): healthy controls (CON group), endotoxemic animals (15mg/kg lipopolysaccharide, LPS group), endotoxemic animals treated with a TLR4 antagonist (1mg/kg CRX-526, LPS+CRX526 group), and controls treated with CRX-526 (C-CRX526 group). Plasma samples were obtained for cytokine measurements at the end of the experiments. Endotoxemia significantly increased leukocyte adhesion in intestinal submucosal venules (e.g., V1 venules: CON 20.4±6.5n/mm(2), LPS 237.5±36.2n/mm(2), p<0.05) and reduced capillary perfusion of the intestinal wall (e.g., longitudinal muscular layer: CON 112.5±5.9cm/cm(2), LPS 71.3±11.0cm/cm(2), p<0.05) at 2h. TLR4 inhibition significantly reduced endotoxemia-associated leukocyte adhesion (V1 venules: 104.3±7.8n/mm(2)) and improved capillary perfusion (longitudinal muscular layer: 111.0±12.3cm/cm(2)). Cytokine release was not significantly affected. The TLR4 pathway may be a target in clinical Gram-negative sepsis since administration of the TLR4 antagonist CRX-526 improved intestinal microcirculation parameters in experimental endotoxemia.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)