Merck
  • Transcriptional coordination and abscisic acid mediated regulation of sucrose transport and sucrose-to-starch metabolism related genes during grain filling in wheat (Triticum aestivum L.).
Transcriptional coordination and abscisic acid mediated regulation of sucrose transport and sucrose-to-starch metabolism related genes during grain filling in wheat (Triticum aestivum L.).
Plant science : an international journal of experimental plant biology (2015-10-18)
Shalini Mukherjee, Aihua Liu, Kirandeep K Deol, Konstanin Kulichikhin, Claudio Stasolla, Anita Brûlé-Babel, Belay T Ayele
ABSTRACT
Combining physiological, molecular and biochemical approaches, this study investigated the transcriptional coordination and abscisic acid (ABA) mediated regulation of genes involved in sucrose import and its conversion to starch during grain filling in wheat. Sucrose import appears to be mediated by seed localized TaSUT1, mainly TaSUT1D, while sucrose cleavage by TaSuSy2. Temporal overlapping of the transcriptional activation of AGPL1 and AGPS1a that encode AGPase with that of the above genes suggests their significance in the synthesis of ADP-glucose; TaAGPL1A and TaAGPL1D contributing the majority of AGPL1 transcripts. ABA induced repressions of TaSUT1, TaSuSy2, TaAGPL1 and TaAGPS1a imply that ABA negatively regulates sucrose import into the endosperm and its subsequent metabolism to ADP-glucose, the substrate for starch synthesis. The formations of amyloses and amylopectin from ADP-glucose appear to be mediated by specific members of GBSS, and SS, SBE and DBE gene families, and the ABA-induced transcriptional change in most of these genes implies that ABA regulates amylose and amylopectin synthesis. The findings provide insights into the molecular mechanisms underlying the coordination and ABA mediated regulation of sucrose transport into the developing endosperm and its subsequent metabolism to starch during grain filling in wheat.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SAFC
Glycine
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%