Merck
  • Copper in Helix pomatia (Gastropoda) is regulated by one single cell type: differently responsive metal pools in rhogocytes.
Copper in Helix pomatia (Gastropoda) is regulated by one single cell type: differently responsive metal pools in rhogocytes.
American journal of physiology. Regulatory, integrative and comparative physiology (2005-05-21)
Reinhard Dallinger, Monika Chabicovsky, Elisabeth Hödl, Caroline Prem, Peter Hunziker, Claudia Manzl
ABSTRACT
Like all other animal species, terrestrial pulmonate snails require Cu as an essential trace element. On the other hand, elevated amounts of Cu can exert toxic effects on snails. The homeostatic regulation of Cu must therefore be a pivotal goal of terrestrial pulmonates to survive. Upon administration of Cu, snails accumulate the metal nearly equally in most of their organs. Quantitative studies in connection with HPLC and electrospray ionization mass spectrometry reveal that a certain fraction of Cu in snails is bound to a Cu-metallothionein (Cu-MT) isoform that occurs in most organs at constant concentrations, irrespective of whether the animals had been exposed to physiological or elevated amounts of Cu. In situ hybridization demonstrates that at the cellular level, the Cu-binding MT isoform is exclusively expressed in the so-called pore cells (or rhogocytes), which can be found in all major snail organs. The number of pore cells with Cu-MT mRNA reaction products remains unaffected by Cu exposure. Rhogocytes also are major storage sites of Cu in a granular form, the metal quickly entering the snail tissues upon elevated exposure. The number of rhogocytes with granular Cu precipitations strongly increases upon Cu administration via food. Thus, whereas Cu-MT in the rhogocytes represents a stable pool of Cu that apparently serves physiological tasks, the granular Cu precipitations form a second, quickly inducible, and more easily available pool of the metal that serves Cu regulation by responding to superphysiological metal exposure.
Product Description
Supelco
5-(4-Dimethylaminobenzylidene)rhodanine, for TLC derivatization, ≥98.0%
Pricing and availability is not currently available.