Merck
  • Rh(I)-catalyzed carbonylative cyclization reactions of alkynes with 2-bromophenylboronic acids leading to indenones.
Rh(I)-catalyzed carbonylative cyclization reactions of alkynes with 2-bromophenylboronic acids leading to indenones.
Journal of the American Chemical Society (2007-04-10)
Yasuyuki Harada, Jun Nakanishi, Hirokazu Fujihara, Mamoru Tobisu, Yoshiya Fukumoto, Naoto Chatani
ABSTRACT
The Rh-catalyzed reaction of alkynes with 2-bromophenylboronic acids involves carbonylative cyclization to give indenones. The key steps in the reaction involve the addition of an arylrhodium(I) species to an alkyne and the oxidative addition of C-Br bonds on the adjacent phenyl ring to give vinylrhodium(I) species II. The regioselectivity depends on both the electronic and the steric nature of the substituents on the alkynes. A bulky group and an electron-withdrawing group favor the -position of indenones. In the case of silyl- or ester-substituted alkynes, the regioselectivity is extremely high. The selectivity increases in the order SiMe3 > COOR > aryl > alkyl. The reaction of norbornene with 2-bromophenylboronic acids under 1 atm of CO gives the corresponding indanone derivative. The reaction of alkynes with 2-bromophenylboronic acids under nitrogen gives naphthalene derivatives, in which two molecules of alkynes are incorporated. A vinylrhodium complex similar to II can also be generated by a different route by employing 2-bromophenyl(trimethylsilyl)acetylene and arylboronic acids in the presence of Rh(I) complex as the catalyst, resulting in the formation of indenones. The reaction of 1-(2-bromophenyl)-hept-2-yn-1-one with PhB(OH)2 in the presence of Rh(I) complex also resulted in carbonylative cyclization to give an indan-1,3-dione derivative.
Sigma-Aldrich
1-(Trimethylsilyl)propyne, 99%
Pricing and availability is not currently available.