Merck
  • Insight into Ion Transfer through the Sub-Nanometer Channels in Zeolitic Imidazolate Frameworks.
Insight into Ion Transfer through the Sub-Nanometer Channels in Zeolitic Imidazolate Frameworks.
Angewandte Chemie (International ed. in English) (2017-03-28)
Ze-Yu Jiang, Hai-Ling Liu, Saud Asif Ahmed, Sumaira Hanif, Shi-Bin Ren, Jing-Juan Xu, Hong-Yuan Chen, Xing-Hua Xia, Kang Wang
ABSTRACT
A crack-free sub-nanometer composite structure for the study of ion transfer was constructed by in situ growth of ZIF-90 [Zn(ICA)
Sigma-Aldrich
2-Imidazolecarboxaldehyde, 97%
Pricing and availability is not currently available.