Merck
  • Patchouli oil ameliorates acute colitis: A targeted metabolite analysis of 2,4,6-trinitrobenzenesulfonic acid-induced rats.
Patchouli oil ameliorates acute colitis: A targeted metabolite analysis of 2,4,6-trinitrobenzenesulfonic acid-induced rats.
Experimental and therapeutic medicine (2017-08-16)
Xiuting Yu, Guanghua Yang, Hua Jiang, Shuhai Lin, Yuhong Liu, Xie Zhang, Huifang Zeng, Ziren Su, Song Huang, Linlin Shen, Xiaojun Zhang
ABSTRACT
The incidence of inflammatory bowel disease (IBD), characterized by chronic, relapsing intestinal inflammation, has continually increased in recent years. A previous study by our group identified five potential metabolic markers possibly associated with the pathology of 2,4,6-trinitrobenzenesulfonic acid (TNBS)-induced IBD in rats. The present study aimed to examine the potential therapeutic effects of the essential oil of
Product Description
Supelco
N-(2-Phenylacetyl)glycine, analytical standard
Pricing and availability is not currently available.