Merck
  • Biological Cleavage of the C–P Bond in Perfluoroalkyl Phosphinic Acids in Male Sprague-Dawley Rats and the Formation of Persistent and Reactive Metabolites.
Biological Cleavage of the C–P Bond in Perfluoroalkyl Phosphinic Acids in Male Sprague-Dawley Rats and the Formation of Persistent and Reactive Metabolites.
Environmental health perspectives (2017-11-15)
Shira Joudan, Leo W Y Yeung, Scott A Mabury
ABSTRACT
Perfluoroalkyl phosphinic acids (PFPiAs) have been detected in humans, wildlife, and various environmental matrices. These compounds have been used with perfluoroalkyl phosphonic acids (PFPAs) as surfactants in consumer products and as nonfoaming additives in pesticide formulations. Unlike the structurally related perfluoroalkyl sulfonic and carboxylic acids, little is known about the biological fate of PFPiAs. We determined the biotransformation products of PFPiAs and some pharmacokinetic parameters in a rat model. Male Sprague-Dawley rats received an oral gavage dose of either C PFPiAs were metabolized to the corresponding PFPAs and 1H-perfluoroalkanes (1H-PFAs), with 70% and 75% biotransformation 2 wk after a single bolus dose for C PFPiAs were metabolized in Sprague-Dawley rats to form persistent PFPAs as well as 1H-PFAs, which contain a labile hydrogen that may undergo further metabolism. These results in rats produced preliminary findings of the pharmacokinetics and metabolism of PFPiAs, which should be further investigated in humans. If there is a parallel between the disposition of these chemicals in humans and rats, then humans with detectable amounts of PFPiAs in their blood may be undergoing continuous exposure. https://doi.org/10.1289/EHP1841.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Pricing and availability is not currently available.