Merck
Modulation of Nitrous Oxide (N
Environmental science & technology (2017-10-31)
Octavio Perez-Garcia, Cody Mankelow, Kartik Chandran, Silas G Villas-Boas, Naresh Singhal
ABSTRACT
Metabolomics provides insights into the actual physiology of cells rather than their mere "potential", as provided by genomic and transcriptomic analysis. We investigate the modulation of nitrous oxide (N
Sigma-Aldrich
Chlorocyclohexane, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl chloroformate, 99%
Pricing and availability is not currently available.