Skip to Content
Merck

860862P

Avanti

6-OH-Cer EOH (d18:1/28:0/18:2)

new

Avanti Research - A Croda Brand

Synonym(s):

N-[28-linoleoyloxy octacosanoyl]-6R-hyxroxy-D-erythro-sphingosine

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C64H121NO6
Molecular Weight:
1000.65
UNSPSC Code:
12352211

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860862P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC[C@@H](O)\C=C\[C@@H](O)[C@H](CO)NC(=O)CCCCCCCCCCCCCCCCCCCCCCCCCCCOC(=O)CCCCCCC\C=C/C\C=C/CCCCC

General description

6-OH-Cer EOH (d18:1/28:0/18:2) is natural component of human skin and is essential in preventing water loss, healing the skin, and maintaining the integrity of the skin.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service