Skip to Content
Merck

CIAH999A9098

ChemScene LLC (Preferred Partner)

7-Fluoro-6-methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

Synonym(s):

7-fluoro-6-methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product CIAH999A9098 is not currently sold in your country.

About This Item

Linear Formula:
C10H13O1N1Cl1F1
CAS Number:
Molecular Weight:
217.67
MDL number:
Product CIAH999A9098 is not currently sold in your country.

Inchi Code

1S/C10H12FNO.ClH/c1-13-10-5-7-2-3-12-6-8(7)4-9(10)11;/h4-5,12H,2-3,6H2,1H3;1H

Inchi Key

JRMGMFQYOPHDTM-UHFFFAOYSA-N

Purity

97%

Salt Data

Hydrochloride

Storage temp.

Store at room temperature, keep dry and cool

Country of Origin

CN

Physical Form

Solid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library