Skip to Content
Merck

ENA024296471

Enamine

4-[(furan-2-yl)methyl]-11-methyl-5-sulfanyl-8-thia-4,6-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),5-trien-3-one

Sign In to View Organizational & Contract Pricing

Select a Size

Product ENA024296471 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
332.45
MDL number:
Product ENA024296471 is not currently sold in your country.

Iupac Name

3-(2-furylmethyl)-7-methyl-2-sulfanyl-5,6,7,8-tetrahydro[1]benzothieno[2,3-d]pyrimidin-4(3H)-one

Inchi Code

1S/C16H16N2O2S2/c1-9-4-5-11-12(7-9)22-14-13(11)15(19)18(16(21)17-14)8-10-3-2-6-20-10/h2-3,6,9H,4-5,7-8H2,1H3,(H,17,21)

InChI key

BJXCEZMOKJVGPI-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-10435

MSDS

https://www.enaminestore.com/catalog/EN300-10435


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library