Skip to Content
Merck

R3877

Rose Bengal sodium salt

Dye content ≥85 %

Synonym(s):

4,5,6,7-Tetrachloro-2′,4′,5′,7′-tetraiodofluorescein sodium salt, Acid Red 94, Bengal Rose B sodium salt

Sign In to View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C20H2Cl4I4Na2O5
CAS Number:
Molecular Weight:
1017.64
Colour Index Number:
45440
Beilstein:
3645857
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

powder

composition

Dye content, ≥85%

solubility

H2O: 1 mg/mL

SMILES string

[Na+].[Na+].[O-]C(=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1C2=C3C=C(I)C(=O)C(I)=C3Oc4c(I)c([O-])c(I)cc24

InChI

1S/C20H4Cl4I4O5.2Na/c21-10-8(9(20(31)32)11(22)13(24)12(10)23)7-3-1-5(25)16(29)14(27)18(3)33-19-4(7)2-6(26)17(30)15(19)28;;/h1-2,29H,(H,31,32);;/q;2*+1/p-2

InChI key

UWBXIFCTIZXXLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maryam Shahi et al.
Lasers in medical science, 36(1), 91-98 (2020-04-17)
The integrity of the structural cerebral cortex is disrupted after stroke either at the macroscopic or microscopic levels. Therefore, many attempts have been gathered to circumvent stroke-associated problems in the brain tissue. The current study was aimed to design an
Matilde Balbi et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 39(8), 1486-1500 (2018-03-10)
Small vessel disease is characterized by sporadic obstruction of small vessels leading to neuronal cell death. These microinfarcts often escape detection by conventional magnetic resonance imaging and are identified only upon postmortem examination. Our work explores a brain-wide microinfarct model
Lun-De Liao et al.
Neurobiology of disease, 75, 53-63 (2015-01-13)
To investigate the potential therapeutic effects of peripheral sensory stimulation during the hyperacute phase of stroke, the present study utilized electrophysiology and photoacoustic imaging techniques to evaluate neural and vascular responses of the rat cortex following ischemic insult. We employed
Matilde Balbi et al.
Cell reports, 34(5), 108696-108696 (2021-02-04)
Alterations in gamma oscillations occur in several neurological disorders, and the entrainment of gamma oscillations has been recently proposed as a treatment for neurodegenerative disease. Optogenetic stimulation enhances recovery in models of stroke when applied weeks after injury; however, the
Bahram Hemmateenejad et al.
Analytical and bioanalytical chemistry, 394(7), 1965-1975 (2009-06-23)
This article describes the use of the net analyte signal (NAS) concept and rank annihilation factor analysis (RAFA) for building two different multivariate standard addition models called "SANAS" and "SARAF." In the former, by the definition of a new subspace

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service