Skip to Content
Merck

V800020

Acetic acid

LR, ≥99.5%

Synonym(s):

Glacial acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

6 X 500 ML

₹2,384.30

4 X 2.5 L

₹5,517.65

₹2,384.30


Estimated to ship on04 February 2026Details


Request a Bulk Order

About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
EC Number:
200-580-7
UNSPSC Code:
12352300
PubChem Substance ID:
Beilstein/REAXYS Number:
506007
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Acetic acid, LR, ≥99.5%

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

SMILES string

[F2C(F2C)13F3C]C(O)=O

grade

LR

vapor density

2.07 (vs air)

product line

Vetec

assay

≥99.5%

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 2

This Item
V800019
grade

LR

grade

AR

assay

≥99.5%

assay

≥99.8%

form

liquid

form

liquid

pH

2.5 (20 °C, 50 g/L)

pH

2.5 (20 °C, 50 g/L)

vapor density

2.07 (vs air)

vapor density

2.07 (vs air)

mp

16.2 °C (lit.)

mp

16.2 °C (lit.)

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

102.2 °F - closed cup

flash_point_c

39 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Taniguchi et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(2), 517-526 (2014-11-12)
Fli-1, a potential predisposing factor for systemic sclerosis (SSc), is constitutively down-regulated in the lesional skin of patients with SSc by an epigenetic mechanism. To investigate the impact of Fli-1 deficiency on the induction of an SSc phenotype in various
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development
Xianbao Deng et al.
The New phytologist, 201(4), 1469-1483 (2013-11-26)
• Chalcone synthase (CHS) is the key enzyme in the first committed step of the flavonoid biosynthetic pathway and catalyzes the stepwise condensation of 4-coumaroyl-CoA and malonyl-CoA to naringenin chalcone. In plants, CHS is often encoded by a small family
N Robichaud et al.
Oncogene, 34(16), 2032-2042 (2014-06-10)
The progression of cancers from primary tumors to invasive and metastatic stages accounts for the overwhelming majority of cancer deaths. Understanding the molecular events which promote metastasis is thus critical in the clinic. Translational control is emerging as an important

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service