Select a Size
€212.00
€382.00
€212.00
About This Item
Skip To
description
Powered by Eupheria Biotech
Quality Level
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CTGCTCAATAGCCAGGCTTCTGGGGACAAAGACATGCAAGCAGGTCTTTCAGTTTGCAGTCAAAGAATCACTTATCCTAAAGCTGCCAACAGATGAGCTCATGAACCCTGCACAGAAGAAGAAAAGAAAACACAGGTTGTGGGCCGCACACTGCAGGAAAATTCAGCTGAAGAAAGATAACAATTCTACACAAGTGTATAACTACCAACCCTGTGACCACCCAGACCGTCCGTGTGACAGCACATGCCCCTGCATCATGACCCAGAACTTTTGTGAAAAGTTCTGCCAGT
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... EZH1(14055), Ezh1(14055)
1 of 4
This Item | EMU014681 | EMU178341 | EMU009031 |
|---|---|---|---|
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Gene Information mouse ... EZH1(14055), Ezh1(14055) | Gene Information mouse ... EZH2(14056), Ezh2(14056) | Gene Information mouse ... H19(14955) | Gene Information mouse ... IGSF8(140559), Igsf8(140559) |
| Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. |
| esiRNA cDNA target sequence CTGCTCAATAGCCAGGCTTCTGGGGACAAAGACATGCAAGCAGGTCTTTCAGTTTGCAGTCAAAGAATCACTTATCCTAAAGCTGCCAACAGATGAGCTCATGAACCCTGCACAGAAGAAGAAAAGAAAACACAGGTTGTGGGCCGCACACTGCAGGAAAATTCAGCTGAAGAAAGATAACAATTCTACACAAGTGTATAACTACCAACCCTGTGACCACCCAGACCGTCCGTGTGACAGCACATGCCCCTGCATCATGACCCAGAACTTTTGTGAAAAGTTCTGCCAGT | esiRNA cDNA target sequence CTGTTCAGAGGGAGCAAAGCTTGCATTCATTTCATACGCTCTTCTGTCGACGATGTTTTAAGTATGACTGCTTCCTACATCCCTTCCATGCAACACCCAACACATATAAGAGGAAGAACACAGAAACAGCTTTGGACAACAAGCCTTGTGGACCACAGTGTTACCAGCATCTGGAGGGAGCTAAGGAGTTTGCTGCTGCTCTTACTGCTGAGCGTATAAAGACACCACCTAAACGCCCAGGGGGGCGCAGAAGAGGAAGACTTCCGAATAACAGTAGCAGACCCAGCACCCCCACCATCAGTGTGCTGGAGTCAAAGGATACAGACAGTGACAGAGAAGCAGGGACTGAAACTGGGGGAGAGAACAATGATAAAGAAGAAGAAGAGAAAAAAGATGAGACGTCCAGCTCCTCTGAAGCAAATTCTCGG | esiRNA cDNA target sequence CAGTGATCGGTGTCTCGAAGAGCTCGGACTGGAGACTAGGCCAGGTCTCCAGCAGAGGTGGATGTGCCTGCCAGTCACTGAAGGCGAGGATGACAGGTGTGGTCAATGTGACAGAAAGACATGACATGGTCCGGTGTGATGGAGAGGACAGAAGGGCAGTCATCCAGCCTTCTTGAACACCATGGGCTGGCGCCTTGTCGTAGAAGCCGTCTGTTCTTTCACTTTTCCCAAAGAGCTAACACTTCTCTGCTGCTCTCTGGATCCTCCTCCCCCTACCTTGAACCCTCAAG | esiRNA cDNA target sequence CCCAGAAGTCATCGGCTAAGACTGCACGGCTTGGGGCCTGAGGATGAAGGCATATACCACTGTGCCCCAAGTGCCTGGGTGCAGCATGCGGACTACAGCTGGTACCAGGCAGGCAGTGCGCGCTCCGGGCCTGTCACAGTCTACCCCTACACGCATGCTGTGGATACCCTATTTGTGCCCCTGTTGGTGGGTACAGGAGTCGCCCTAGTTACCGGCGCCAGCGTCCTGGCTACCATCACCTGCTGCTTTATGAAGAGGATGCGGAAGCGGTGATCTGTAGCACCCCAGTTCTTGCAGGTGTTGACTGTCTTCCAGCCAGTGCCACCCCTCCTCTGGTTGCCTGGATACCCTCTCCCTCTGTCCAGCCCACCTTTAATTTATTTGACCAGCCATCACCTAGAGTGGGAGACATGGC |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service