Skip to Content
Merck

FLUH99C88B4D

Fluorochem (Preferred Partner)

5-Bromo-7-fluoro-2,3-dihydro-1H-inden-1-one

Sign In to View Organizational & Contract Pricing

Select a Size

250 MG

¥10,000

1 G

¥11,300

5 G

¥23,000

10 G

¥37,700

25 G

¥76,600

Vendor SKU: F238513-250MG

¥10,000


Ships in 15 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

5-bromo-7-fluoro-1-indanone

Inchi Code

1S/C9H6BrFO/c10-6-3-5-1-2-8(12)9(5)7(11)4-6/h3-4H,1-2H2

InChI key

VMPKZIKGIGAVGK-UHFFFAOYSA-N

storage temp.

Ambient Storage

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://fluorochem.co.uk/product/F238513

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://sds.fluorochem.co.uk/eng/sds_F238513_en.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service