Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207
Pricing and availability is not currently available.
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
ATCCTTCCCCAGCTGAAACTTCACTTGAGACCTTCTCATTGGCCATCATCGACATCTGGATTCATGATACAGAGAAACTTGAGTAATGATGACAATAGCAGTCCAGCCTGCATGGATGCTGACATGGATGCTGTCTCCCATTCTTACCACTGGATGTCAAGAACTGCTAGATGCGATCCAGCCAGTCTGTACTCGACTCTCAGTTCCATCTTCTCAACTCACAGTGTCTTCCAAAGTCCATGCCACACCACAGCCTGGAAGATC
GenBank accession no.
shipped in
ambient
storage temp.
−20°C
General description
MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
GenBank is a registered trademark of United States Department of Health and Human Services
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.