Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GCACATCAAAGAAGCCCATTGAACCCCAAGTAGGATGAATGCAAAAAGATCTACCCAGGTATATCAGAGTAAGAATGCTGAAATCTAAAGACAGAAAATACTGCAAGCAGCAAGTGGAAAATGACTTTGCTTGGACACAGGAAACTTAGTGAGATTAAGATCTTCTCATCAGGAATAATGGCGACCAGGAAAGAGTGGGGTGACATATTCAAAGTACTGAAAGAAGAAATAAAAAACAAAAAAAAGCTGTCAACCAAGAATTATACATCCAGCAAAACTATCTGTAAAAATG
GenBank accession no.
shipped in
ambient
storage temp.
−20°C
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting human RP11-298H24.1 | description targeting human AL358334 | description targeting human RP11-319F12.2 | description targeting human AL136307 |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| esiRNA cDNA target sequence GCACATCAAAGAAGCCCATTGAACCCCAAGTAGGATGAATGCAAAAAGATCTACCCAGGTATATCAGAGTAAGAATGCTGAAATCTAAAGACAGAAAATACTGCAAGCAGCAAGTGGAAAATGACTTTGCTTGGACACAGGAAACTTAGTGAGATTAAGATCTTCTCATCAGGAATAATGGCGACCAGGAAAGAGTGGGGTGACATATTCAAAGTACTGAAAGAAGAAATAAAAAACAAAAAAAAGCTGTCAACCAAGAATTATACATCCAGCAAAACTATCTGTAAAAATG | esiRNA cDNA target sequence AGCATTTGTTCCGCCTCTCCCCTCTAGAGTGTGGAGACAGGAGGACGTCTGTGTAGGGAAGCACGTCGTTGGCAGACTGCTGTTTTGCCGATGTTTATTTGTTATGCCATTATGACTGTTAGGATTTCTCACCTGGAGTGGCTTCCGCCATAAGCGCCGGGATCTGGAAGGACGGCACGTTTGACTTCTGTTGTCCCAACCTCACTTGCCAGCCCTTGGCACCGCCCCAGCTTGGATCCTCAGGGCGCCTCTGTTATTTTCATTCATTTTTTTTGAGGACTCCAGGGTTCAGGTTTCTTAACTCCTGGACCAAAGTTCGCCCCGCTCGTAGCACCCTGTCTCTCATCCTTAGAGATTCTGAAGTTTGGACTTGAGCTTTTGGGGCAGAGACTGAATTGGGCCTTATGCAACTCTTGCAATACCGATGACCTGCAAAGGACAGTATGGAAGAGATGAGCATTAGAGTTTGGATCCGGATGG | esiRNA cDNA target sequence AGAGCACCAAAAGCACCAGTGTGCAGAAATCAGAGACCTCAAGCTAAAAGTCATAAAAGGACAAGAACCTGGAAGTCGTCGAACTGTGGAAGCAAAAGACCTGTAATGCTCCTGCTTGCCGAGCTGTGAGCAGTGGTAGTAAAAGAGCAGTAACATTCTCTCCTGCTTGCCAAACAGGAGAGAAGCCACTGGCACCACTCCCTCCCACTCACTGAACTATAGAAGAGAAAAAATTGAAACACCAAAATTGGGGAA | esiRNA cDNA target sequence GTAAAGCCGTTTGCACCACTATTATCCTTCTGCTCCTTTTAAAACCTAACAGGCAGGCCCAACTTGGAAACTTCCAAGAATGAAACACCACCTTCAGAAAACGTGGTAATTGATAGAAGAGTCTGGTGCTACTGCATGATGCATTCCATACCCTCTGGGCTCCTCTCTCTAGTTCTTTAACCTAGTCTCTCTCAAGCTATGATGAGAAAGGCACTGATTATTTCAAAATGGTGCAAATCACCTGCTGTTTAAAAAACAAGATAAATCCCCCAAAAACGAAAAAAATGAAGTTGTCAAAGACACTGGAATGAAATTTCTGAAGAACCAATCTTTTCCCACTGTGTTTCTCCATCAAAAGGAAAAGCTAGGCAAGCTTGTTGAAATGATCAGCTCCATGTGTTCAGCCTGAGTATAGGGTAGCAGGAAACTGCACCCTAATTTTAGTGTTCTGCATGGACTCTCATTACTGCCCAAGCACATGGCTCTCACATGCATAAGTCCTTCTTGAGTACTGGTTGCTAACCCTCCTCCACACTTGG |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| GenBank accession no. | GenBank accession no. | GenBank accession no. | GenBank accession no. |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
