Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CAGAAATGCCAAAGCACTCAAAGCAAAGAAAGAAGATGGGAATAAAGCATTTAAGGAAGGAAATTACAAACTAGCATATGAACTGTACACAGAAGCCCTGGGGATAGACCCCAACAATATAAAAACAAATGCTAAACTCTACTGTAATCGGGGTACGGTTAATTCCAAGCTTAGGAAACTAGATGATGCAATAGAAGACTGCACAAATGCAGTGAAGCTTGATGACACTTACATAAAAGCCTACTTGAGAAGAGCTCAGTGTTACATGGACACAGAACAGTATGAAGAAGCAGTACGAGACTATGAAAAAGTATACCAGACAGAGAAAACAAAAGAACACAAACAGCTCCTAAAAAATGCGCAGCTGGAACTGAAGAAGAGTAAGAGGAAAGATTACTACAAGATTCTAGGAGTGGACAAGAATGCCTCTGAGGACGAGATCAAGAAAGCTTATCGGAAACGGGCCTTGATGCACCATCCAGATCGGCATAGTGGAGCCAGTGCTGA
Ensembl | human accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... DNAJC7(7266)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting human DNAJC7 | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description targeting mouse Dnajc7 |
| product line MISSION® | product line Prestige Antibodies® Powered by Atlas Antibodies | product line Prestige Antibodies® Powered by Atlas Antibodies | product line MISSION® |
| Ensembl | human accession no. | Ensembl | human accession no. - | Ensembl | human accession no. - | Ensembl | human accession no. - |
| Gene Information human ... DNAJC7(7266) | Gene Information human ... DNAJC7(7266) | Gene Information human ... DNAJC7(7266) | Gene Information mouse ... DNAJC7(56354), Dnajc7(56354) |
| esiRNA cDNA target sequence CAGAAATGCCAAAGCACTCAAAGCAAAGAAAGAAGATGGGAATAAAGCATTTAAGGAAGGAAATTACAAACTAGCATATGAACTGTACACAGAAGCCCTGGGGATAGACCCCAACAATATAAAAACAAATGCTAAACTCTACTGTAATCGGGGTACGGTTAATTCCAAGCTTAGGAAACTAGATGATGCAATAGAAGACTGCACAAATGCAGTGAAGCTTGATGACACTTACATAAAAGCCTACTTGAGAAGAGCTCAGTGTTACATGGACACAGAACAGTATGAAGAAGCAGTACGAGACTATGAAAAAGTATACCAGACAGAGAAAACAAAAGAACACAAACAGCTCCTAAAAAATGCGCAGCTGGAACTGAAGAAGAGTAAGAGGAAAGATTACTACAAGATTCTAGGAGTGGACAAGAATGCCTCTGAGGACGAGATCAAGAAAGCTTATCGGAAACGGGCCTTGATGCACCATCCAGATCGGCATAGTGGAGCCAGTGCTGA | esiRNA cDNA target sequence - | esiRNA cDNA target sequence - | esiRNA cDNA target sequence AGAAGGCTTGTGTCGCTTGTAGAAATGCCAAAGCCCTTAAAGCCAAGAAGGAAGATGGGAATAAAGCCTTTAAGGAAGGAAATTACAAGCTAGCATATGAACTGTACACAGAAGCCTTGGGGATAGATCCCAACAACATAAAAACAAATGCTAAACTCTACTGTAATCGGGGTACGGTTAATTCCAAGCTTAGGCAACTGGAAGATGCCATAGAAGACTGTACAAATGCGGTGAAGCTCGATGACACTTACATCAAAGCCTACCTGAGAAGAGCTCAGTGTTACATGGACACAGAGCAGTTTGAAGAAGCCGTGCGGGACTATGAAAAAGTGTATCAGACGGAGAAAACAAAAGAACACAAACAGCTCCTTAAGAATGCACAGCTGGAACTGAAGAAGAGCAAGAGGAAAGATTACTACAAGATCCTGGGAGTGGACAAGAATGCCTCTGAGGACGAGATCAAGAAAGCTTACCGGAAACGGGCCTTGATGCACCATCCAGATCGGCACAGTGGGGCCAGTGCCGAAGTTCAGAAGGA |
| form lyophilized powder | form buffered aqueous glycerol solution | form buffered aqueous glycerol solution | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


