Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CTACAATGCACGTTGGGATGCATATAGAATGCATAACATCGTTTCACTTACGTGAAAGGCACTAATGACTGCTCCACAAAAATTGTCTGCAGAGAAAAACTTGAGGCTGAACTCATGGCTGAGTGCTTTGGTTATATGAACAAGAAATGTCTGAAGTAGGAAGAAATGATATGAGTAACACACACACACACACACACACACACACACACACACACCTACTTACTATAGATAGGGAAGTCATGACAGATCAATATAAGCAGTCTTCCAAAGTCCAGC
GenBank accession no.
shipped in
ambient
storage temp.
−20°C
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting mouse Ak034429 | description targeting human RP5-1069C8.3 | description targeting human AC023137.2 | description targeting human AC079766 |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| esiRNA cDNA target sequence CTACAATGCACGTTGGGATGCATATAGAATGCATAACATCGTTTCACTTACGTGAAAGGCACTAATGACTGCTCCACAAAAATTGTCTGCAGAGAAAAACTTGAGGCTGAACTCATGGCTGAGTGCTTTGGTTATATGAACAAGAAATGTCTGAAGTAGGAAGAAATGATATGAGTAACACACACACACACACACACACACACACACACACACACCTACTTACTATAGATAGGGAAGTCATGACAGATCAATATAAGCAGTCTTCCAAAGTCCAGC | esiRNA cDNA target sequence TTGGCCTACCTGTCAGTCCTGCCCTGCGTGTCACCTCAGCCTGGAAGTCCATAGGACTGTGATGAAGATGACTGCTTGCATTTTCTGAATCACTTTGTCGATGAGACATGCATCCTATTAGATCTAACAAAGCTTCACAGAAAGGTTGTTTTGAAGTTGACACTGAGGAAGATATAAGTGCTCTAGGATCAGTATGTGAGGCTGCCACAAACATGCTATAGTCTGCACATTCTTGCAGAGAGGTTGCAGAGTCTGCTTGGGAGAGAAGCCTCAAAAACATGGCTGGTGTCTTGGCAGAGTTTGCTCTGGCCCCCAGGAGGGTGTACAAACAGCCTTCATGAGAAGGTGACTTGCTGGCCACTCTTAACCACTAGAAGAAACTGATGCCATGGAACTTTGAAAAATCAGAAAGCAATTCCACAAAAGAAGAACCTCTGAAGATAGTAGAGCCCAATGAAGATAAATGACATCGCCCATAGGAGGGAACAGTGGTATCAACCTAAAGGGGCTCACTG | esiRNA cDNA target sequence ATAATCCTGCCGGAACACACTCTTCTTGCTGCATTAGGATCCAACTGGAAAACATAAAGATGAATTGCTGTAGGCTGGAAAATCAATTAGGAAAAAGAGGGATGTGATTCAGACACAGGACTAGCTTTGTGATCCCTGTGCTCCGGAGAAACCTCCACCCTGGCCTCAAGGAGGCTGTGAAACCAAAAGGGACCTTGGGTGCCAGCTTGCTGGGTCCTTACTTACTTGGTCCATCTTCTGGCTGGAAGGAGCACCTGGGCTCTCAGAGTGCCCCCCGACTCTCAGCAGCTCCTTT | esiRNA cDNA target sequence GTTTACGGCGGCAAGAGAGCTCAATTAATTTCTGAGGGAGGACGGCAGGATGCGACCCCTTGTGCGTGGCCTAGGAGGAACTGTCCCTGTGTGAATTTATGTTCTGGTAATTTAAAAAGATTTATATTAAGCTTTTCTTTCATTTTGAGAGTAGATTAAAATTCTAATGGAAACCATTTGTATGCATTTGTTGAATTGAAGAGTATTTATGGCTGTGTAAAAAAAATGCTGCAAAATTCTGCCGATAATAAAGTCAGTGAAGCAAGAATAAGTGTAAAAACAGGAAGGAAATACGCGA |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| GenBank accession no. | GenBank accession no. | GenBank accession no. | GenBank accession no. |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
